Learn More
Invitrogen™ GlycoBlue™ Coprecipitant (15 mg/mL)

Description
GlycoBlue Coprecipitant consists of a blue dye covalently linked to glycogen, a branched chain carbohydrate, which is useful as a nucleic acid coprecipitant. The attached dye increases visibility of the pellet. This product is an ideal coprecipitant in nuclease protection assays at 1/100 dilution of stock solution.
Features of GlycoBlue™ Coprecipitant:
• Ideal for RT-PCR
• Increases pellet mass and visibility
• Quantitative recovery of low concentrations (ng/mL) of nucleic acid
• Prevents pellet loss in nuclease protection assays
Order Info
Shipping Condition: Dry Ice

Specifications
Specifications
Concentration | 15 mg/mL |
Content And Storage | Store at –20°C. |
Packaging Type | Tube |
Product Line | Ambion™, GlycoBlue™ |
Purity | Molecular Biology Grade |
Quantity | 300 μL |
Shipping Condition | Dry Ice |
Form | Liquid |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.