Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Glycogen, From Oyster

Catalog No. AC422950050
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle

Glycogen, CAS # 9005-79-2 is a highly branched, polysaccharide form of glucose used for energy storage in humans and other animals. | CAS: 9005-79-2 | C24H42O21 | 666.579 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Glycogen is the main form in which energy is stored in animals as well as other higher organisms
  • It consists of a core protein unit surrounded by highly branched glucose units
  • Glycogen is synthesized through a series of enzymatic reactions and stored mainly in the liver and skeletal muscle to provide energy during fasting periods and during muscle contraction
  • Glycogen is an analog of starch, an energy store in plants, though it is both more compact and more highly branched than starch

Applications

  • Glycogen is most commonly used as an inert carrier substance for nucleic acid precipitation that may increase DNA and RNA extraction efficiency
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9005-79-2
Molecular Formula C24H42O21
Molecular Weight (g/mol) 666.579
MDL Number MFCD00081547
InChI Key BYSGBSNPRWKUQH-UJDJLXLFSA-N
Synonym glycogen, 2r,3r,4s,5s,6r-2-2r,3s,4r,5r,6r-4,5-dihydroxy-6-2r,3s,4r,5r,6s-4,5,6-trihydroxy-2-hydroxymethyl oxan-3-yl oxy-2-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy methyl oxan-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, glycogen from bovine liver, 2r,3r,4s,5s,6r-2-2r,3s,4r,5r,6r-4,5-dihydroxy-6-2r,3s,4r,5r,6s-4,5,6-trihydroxy-2-hydroxymethyl oxan-3-yl oxy-2-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxymethyl oxan-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, 2s,3r,4s,5s,6r-2-2r,3s,4r,5r,6r-4,5-dihydroxy-3-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy-6-2r,3s,4r,5r,6s-4,5,6-trihydroxy-2-hydroxymethyl oxan-3-yl oxyoxan-2-yl methoxy-6-hydroxymethyl oxane-3,4,5-triol, 4-o-4-o-alpha-d-glucopyranosyl-6-o-alpha-d-glucopyranosyl-alpha-d-glucopyranosyl-alpha-d-glucopyranose
PubChem CID 439177
ChEBI CHEBI:28087
IUPAC Name (2S,3R,4S,5S,6R)-2-[[(2R,3S,4R,5R,6R)-4,5-dihydroxy-3-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-2-yl]methoxy]-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3C(OC(C(C3O)O)O)CO)O)O)OC4C(C(C(C(O4)CO)O)O)O)O)O)O)O

Specifications

Color Beige to White
Loss on Drying 10% max. (105°C, 3 hrs)
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula (C6H10O5)n
Quantity 5 g
Merck Index 15, 4531
Specific Rotation Condition +175° (20°C c=0.1,water)
Specific Rotation +160.0° to +180.0° (20°C, 589nm) (c=0.1, water)
Physical Form Powder
Chemical Name or Material Glycogen, From oyster
Show More Show Less
EINECSNumber 232-683-8
RTECSNumber MC2700000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.