Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glycogen phosphorylase, muscle form Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297991AF594
Description
Glycogen phosphorylase, muscle form Polyclonal specifically detects Glycogen phosphorylase, muscle form in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Glycogen phosphorylase, muscle form | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
EC 2.4.1.1, glycogen phosphorylase, muscle form, myophosphorylase, phosphorylase, glycogen, muscle, phosphorylase, glycogen; muscle | |
Produced in rabbits immunized with E. coli-derived Human Glycogen phosphorylase, muscle form fragment. | |
0.1 mL | |
GPCR | |
5837 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction