Learn More
Glycogen Synthase Kinase 3b (GSK3b) Polyclonal Antibody, Abbomax
Rabbit Polyclonal Antibody
Supplier: Abbomax Inc 5007614

Description
Positive control: Mouse muscle tissue lysate Cellular location: Cytoplasmic.
Glycogen Synthase Kinase 3b (GSK3b) Polyclonal antibody specifically detects Glycogen Synthase Kinase 3b (GSK3b) in Bovine, Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
Glycogen Synthase Kinase 3b (GSK3b) | |
Polyclonal | |
Unconjugated | |
GSK3B | |
7330414F15Rik; 8430431H08Rik; C86142; EC 2.7.11.26; FA; Factor A; fb68h05; fk80d11; glycogen synthase kinase 3; glycogen synthase kinase 3 beta; glycogen synthase kinase 3 beta L homeolog; glycogen synthase kinase 3 beta variant 1; glycogen synthase kinase 3 beta variant 2; glycogen synthase kinase 3 beta variant 3; glycogen synthase kinase 3 beta variant 4; glycogen synthase kinase 3 beta variant 5; glycogen synthase kinase 3 beta, genome duplicate a; glycogen synthase kinase-3 beta; glycogen synthase kinase-3 beta splice variant X1; glycogen synthase kinase-3 beta splice variant X2; glycogen synthase kinase-3 beta splice variant X3; GSK 3 beta; GSK 3B; gsk3; GSK-3; GSK3 beta; GSK-3 beta; GSK-3 beta GSK3B; GSK-3[b]; Gsk3b; GSK-3b; gsk3b.L; gsk3ba; gsk3beta; GSK-3beta; gsk3-beta; GSK3beta isoform; intracellular kinase; pp37; serine/threonine-protein kinase GSK3B; wu:fb68h05; wu:fk80d11; XELAEV_180112401mg; xgsk-3; Xgsk-3 protein | |
A synthetic peptide corresponding to the N-terminus of GSK-3 beta protein | |
100 μg | |
Primary | |
Bovine, Human, Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with proprietary stabilizer and 0.01% sodium azide | |
P18266, P49841, Q9WV60 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2932, 56637, 790875, 84027 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.