Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glycogen Synthase, p Ser641 Antibody (1D1), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Glycogen Synthase |
---|---|
Clone | 1D1 |
Dilution | Western Blot 1:500-1:5000, ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200 |
Applications | Western Blot, ELISA, Immunocytochemistry |
Classification | Monoclonal |
Description
Glycogen Synthase Monoclonal antibody specifically detects Glycogen Synthase in Human samples. It is validated for Western Blot, ELISA, ImmunocytochemistrySpecifications
Glycogen Synthase | |
Western Blot 1:500-1:5000, ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
EC 2.4.1.11, glycogen [starch] synthase, muscle, glycogen synthase 1 (muscle), GYSGSY | |
A synthesized peptide derived from Human Glycogen Synthase [p Ser641] [UniProt P13807] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
1D1 | |
Western Blot, ELISA, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Phospho Specific | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
2997 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title