Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycolic Acid, 70%, High Purity, Spectrum™ Chemical
SDP

Catalog No. 18603028
Click to view available options
Quantity:
100 mL
500 mL
4 L
6 x 500 mL
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum

G1059, 79-14-1, C2H4O3

Spectrum™ Chemical Glycolic Acid, 70 Percent, High Purity , is slightly stronger than acetic acid. It is often included into emulsion polymers, solvents and additives for ink and paint in order to improve flow properties and impart gloss. It is used in surface treatment products that increase the coefficient of friction on tile flooring. Glycolic acid is a useful synthetic organic intermediate for oxidation-reduction, esterification and long chain polymerizations.

Chemical Identifiers

CAS 79-14-1, 7732-18-5
Molecular Formula C2H4O3
Molecular Weight (g/mol) 76.05
InChI Key AEMRFAOFKBGASW-UHFFFAOYSA-N
IUPAC Name 2-hydroxyacetic acid
SMILES OCC(O)=O

Specifications

CAS Max % 70%, 30%
UN Number UN3265
Grade High Purity
Quantity 4 L
Packaging Amber Glass Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.