Learn More
Thermo Scientific™ GlycoLink™ Coupling Catalyst PROMO
Description
The GlycoLink Coupling Catalyst is a two-component kit composed of aniline and sodium acetate buffer. The catalyst accelerates the coupling rate of hydrazide and alkoxyamine moieties with reactive aldehydes that have been generated by periodate-oxidation of cis-diols in glycoproteins. The GlycoLink Coupling Catalyst increases the coupling efficiency of these reactions, decreasing the molar excess amounts of labeling reagents that are required for conjugation reactions. The catalyst is also useful for increasing the binding efficiency of oxidized glycoproteins or other carbohydrate-containing molecules to hydrazide resins.
Highlights:
Optimized – catalyzes aldehyde-hydrazide and aldehyde-alkoxyamine conjugation reaction
Efficient – maximizes coupling efficiency and decreases required coupling reaction incubation time
Gentle – does not interfere with protein function and is easily removed by desalting or dialysis
Recommended for:
Labeling aldehyde-containing molecules with alkoxyamine-biotins; Labeling aldehyde-containing molecules with hydrazide-biotins; Increase coupling efficiency of aldehyde-containing molecules to hydrazide resin
Specifications
Specifications
| Content And Storage | Store at 4°C. |
| Product Line | GlycoLink |
| Form | Liquid |
| Quantity | 100 mL |
| Product Type | Coupling Catalyst |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.