Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycolonitrile (ca. 52% in Water, ca. 9.7mol/L) (stabilized with H2SO4), TCI America™
SDP

Catalog No. h019125g
Click to view available options
Quantity:
25 g

Chemical Identifiers

CAS 107-16-4
Molecular Formula C2H3NO
Molecular Weight (g/mol) 57.05
MDL Number MFCD00042731
InChI Key LTYRAPJYLUPLCI-UHFFFAOYSA-N
Synonym Formaldehyde Cyanhydrin, Hydroxyacetonitrile
PubChem CID 7857
IUPAC Name 2-hydroxyacetonitrile
SMILES OCC#N

Specifications

Melting Point -67°C
Color Yellow
Boiling Point 183°C
Quantity 25 g
UN Number 3276
Formula Weight 57.05
Physical Form Liquid
Chemical Name or Material Glycolonitrile (ca. 52% in Water, ca. 9.7mol/L) (stabilized with H2SO4)
EINECSNumber (9)-0385
RTECSNumber AM0350000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.