Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycophorin A Ab-4 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS1843PABX Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS1843PABX 200 μL Concentrated 1mg/mL
MS1843P1ABX 100 μL Concentrated 1mg/mL
MS1843P1 500 μL Concentrated 200μg/mL
MS1843P 1 mL Concentrated 200μg/mL
MS1843R7 7 mL Prediluted -
MS1843P0 100 μL Concentrated 200μg/mL
MS1843R1 1 mL Prediluted -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. MS1843PABX Supplier Epredia Supplier No. MS1843PABX
Add to Cart
Add to Cart

Epredia™ Glycophorin A Ab-4, Mouse Monoclonal Antibody provides accurate, reproducible results.

Glycophorin A is a sialoglycoprotein present on red blood cells (RBC) and erythroid precursor cells. It has been shown that glycophorins acts as the receptor for Sandei virus and parvovirus.

Host Species: Mouse

Clone: JC159

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Extracellular domain

Molecular Weight: 39 kDa

Positive Control: Human erythrocytes in placenta

Cellular Localization: Membrane Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Glycophorin A Ab-4
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone JC159
Concentration 1mg/mL
Conjugate Unconjugated
Host Species Mouse
Quantity 200 μL
Regulatory Status IVD
Research Discipline Infectious Disease
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.