Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glycophorin A Antibody (GYPA/3219R), Alexa Fluor™ 750, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308261AF750
Description
Glycophorin A Monoclonal antibody specifically detects Glycophorin A in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Glycophorin A | |
Monoclonal | |
Alexa Fluor 750 | |
CD235a antigen, glycophorin A (includes MN blood group), glycophorin A (MNS blood group), glycophorin Erik, glycophorin MiI, glycophorin MiIII, glycophorin MiV, glycophorin MiX, glycophorin SAT, glycophorin Sta type C, glycophorin-A, GPA, GPErik, GpMiIII, GPSAT, HGpMiIII, HGpMiV, HGpMiX, HGpMiXI, HGpSta(C), Mi.V glycoprotein (24 AA), MN sialoglycoprotein, MNS, PAS-2, recombinant glycophorin A-B Miltenberger-DR | |
Recombinant full-length human Glycophorin A protein (Uniprot: P02724) | |
100 μg | |
Cancer, Signal Transduction | |
2993 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
GYPA/3219R | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction