Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glycophorin A Antibody (M2A1), PerCP, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP281107PCP
Description
Glycophorin A Monoclonal antibody specifically detects Glycophorin A in Human samples. It is validated for ELISA, Immunoblot, Functional Assay (Inhibition)Specifications
| Glycophorin A | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunoblot, Functional Inhibition Assay | |
| M2A1 | |
| ELISA, Immunoblotting, Functional (Inhibition) | |
| CD235a antigen, glycophorin A (includes MN blood group), glycophorin A (MNS blood group), glycophorin Erik, glycophorin MiI, glycophorin MiIII, glycophorin MiV, glycophorin MiX, glycophorin SAT, glycophorin Sta type C, glycophorin-A, GPA, GPErik, GpMiIII, GPSAT, HGpMiIII, HGpMiV, HGpMiX, HGpMiXI, HGpSta(C), Mi.V glycoprotein (24 AA), MN sialoglycoprotein, MNS, PAS-2, recombinant glycophorin A-B Miltenberger-DR | |
| M2A1 was prepared by immunzing BALB/c mice with human red blood cells of the M phenotype, and was screened by testing for anti-M activity. | |
| 0.1 mL | |
| Cancer, Signal Transduction | |
| 2993 | |
| Store at 4C in the dark. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction