Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glycophorin A Antibody (YTH89.1) - BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
$206.50 - $499.50
Specifications
Antigen | Glycophorin A |
---|---|
Clone | YTH89.1 |
Concentration | 1.0 mg/mL |
Dilution | Flow Cytometry 1:10-1:25, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:1000, Immunohistochemistry-Frozen 1:100-1:1000 |
Applications | Immunohistochemistry (Frozen), Immunohistochemistry, Flow Cytometry, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017681
![]() |
Novus Biologicals
NB100650150.025MG |
0.025 mg |
Each for $206.50
|
|
|||||
NB10065015
![]() |
Novus Biologicals
NB10065015 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
Glycophorin A Monoclonal specifically detects Glycophorin A in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Glycophorin A | |
1.0 mg/mL | |
Immunohistochemistry (Frozen), Immunohistochemistry, Flow Cytometry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rat | |
Cancer, Signal Transduction | |
P02724 | |
2993 | |
Made to Human CD235a | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
YTH89.1 | |
Flow Cytometry 1:10-1:25, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:1000, Immunohistochemistry-Frozen 1:100-1:1000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD235a antigen, glycophorin A (includes MN blood group), glycophorin A (MNS blood group), glycophorin Erik, glycophorin MiI, glycophorin MiIII, glycophorin MiV, glycophorin MiX, glycophorin SAT, glycophorin Sta type C, glycophorin-A, GPA, GPErik, GpMiIII, GPSAT, HGpMiIII, HGpMiV, HGpMiX, HGpMiXI, HGpSta(C), Mi.V glycoprotein (24 AA), MN sialoglycoprotein, MNS, PAS-2, recombinant glycophorin A-B Miltenberger-DR | |
GYPA | |
IgG2b | |
Protein G purified | |
Recognizes glycophorin A, a major sialoglycoprotein of the human erythrocyte membrane. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title