Learn More
Glycophorin A Antibody (YTH89.1), Janelia Fluor™ 646, Novus Biologicals™
SDP
Rat Monoclonal Antibody
Supplier:  Novus Biologicals NB10065015JF646
Catalog No. NB065015646
Only null left
Add to Cart
Description
Description
Glycophorin A Monoclonal specifically detects Glycophorin A in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
Specifications
GlycophorinA
Monoclonal
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
GYPA
Made to Human CD235a
0.1 mL
Primary
Recognizes glycophorin A, a major sialoglycoprotein of the human erythrocyte membrane.
Store at 4°C in the dark.
IgG2b
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
YTH89.1
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
CD235a antigen, glycophorin A (includes MN blood group), glycophorin A (MNS blood group), glycophorin Erik, glycophorin MiI, glycophorin MiIII, glycophorin MiV, glycophorin MiX, glycophorin SAT, glycophorin Sta type C, glycophorin-A, GPA, GPErik, GpMiIII, GPSAT, HGpMiIII, HGpMiV, HGpMiX, HGpMiXI, HGpSta(C), Mi.V glycoprotein (24 AA), MN sialoglycoprotein, MNS, PAS-2, recombinant glycophorin A-B Miltenberger-DR
Rat
Protein G purified
Cancer, Signal Transduction
2993
Human
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.