Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycophorin A Mouse anti-Human, R-PE, Clone: CMRF14, Southern Biotech™
SDP

Catalog No. OB986109 Shop All Southern Biotechnology Associates Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OB986109 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OB986109 Supplier Southern Biotechnology Associates Supplier No. 986109
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SouthernBiotech anti-human primary antibodies are available in a variety of conjugated formats including Alexa Fluor and Pacific Blue.

Specifications

Antigen Glycophorin A
Applications Flow Cytometry
Classification Monoclonal
Clone CMRF14
Concentration Lot specific
Conjugate R-PE
Formulation Supplied in PBS/NaN3 and a stabilizing agent
Host Species Mouse
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 2-8°C; Protect from light; Do not freeze
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.