Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glycophorin A Mouse, Clone: GPHN02, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2477140.2MG
Description
Glycophorin A Monoclonal antibody specifically detects Antigen in Bovine, Human samples. It is validated for Agglutination, Flow Cytometry, Immunofluorescence.Specifications
Glycophorin A | |
Monoclonal | |
1.0 mg/mL | |
Flow Cytometry : 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence : 0.5 - 1.0 ug/ml, Erythrocyte Agglutination, CyTOF-ready | |
CD235a antigen, glycophorin A (includes MN blood group), glycophorin A (MNS blood group), glycophorin Erik, glycophorin MiI, glycophorin MiIII, glycophorin MiV, glycophorin MiX, glycophorin SAT, glycophorin Sta type C, glycophorin-A, GPA, GPErik, GpMiIII, GPSAT, HGpMiIII, HGpMiV, HGpMiX, HGpMiXI, HGpSta(C), Mi.V glycoprotein (24 AA), MN sialoglycoprotein, MNS, PAS-2, recombinant glycophorin A-B Miltenberger-DR | |
Mouse | |
39 kDa | |
0.2 mg | |
Cancer, Signal Transduction | |
2993 | |
Human, Bovine | |
Purified |
Flow Cytometry, Immunofluorescence, Agglutination, CyTOF | |
GPHN02 | |
Unconjugated | |
PBS with No Preservative | |
GYPA | |
Human erythrocytes treated with neuraminidase | |
Protein A or G purified | |
RUO | |
Primary | |
Recognizes a sialoglycoprotein of 39kDa, identified as glycophorin A (GPA). It is present on red blood cells (RBC) and erythroid precursor cells. It has been shown that glycophorin acts as the receptor for Sandei virus and parvovirus. Glycophorins A (GPA) and B (GPB), which are single, trans-membrane sialoglycoproteins. GPA is the carrier of blood group M and N specificities, while GPB accounts for S and U specificities. GPA and GPB provide the cells with a large mucin like surface and it has been suggested this provides a barrier to cell fusion, so minimizing aggregation between red blood cells in the circulation. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgM κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction