Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glycophorin A Rabbit anti-Human, Clone: GYPA/3219R, Novus Biologicals™

Rabbit Monoclonal Antibody
$304.50
Specifications
| Antigen | Glycophorin A |
|---|---|
| Clone | GYPA/3219R |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 0.1-0.2 ug/ml |
| Applications | Immunohistochemistry (Paraffin) |
Description
Glycophorin A Monoclonal specifically detects Glycophorin A in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
| Glycophorin A | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Signal Transduction | |
| 10 mM PBS with 0.05% BSA | |
| 2993 | |
| IgG | |
| Protein A or G purified |
| GYPA/3219R | |
| Immunohistochemistry-Paraffin 0.1-0.2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD235a antigen, glycophorin A (includes MN blood group), glycophorin A (MNS blood group), glycophorin Erik, glycophorin MiI, glycophorin MiIII, glycophorin MiV, glycophorin MiX, glycophorin SAT, glycophorin Sta type C, glycophorin-A, GPA, GPErik, GpMiIII, GPSAT, HGpMiIII, HGpMiV, HGpMiX, HGpMiXI, HGpSta(C), Mi.V glycoprotein (24 AA), MN sialoglycoprotein, MNS, PAS-2, recombinant glycophorin A-B Miltenberger-DR | |
| Recombinant full-length human Glycophorin A protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title