Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glycophorin C Antibody (BRIC10), Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $499.50
Specifications
| Antigen | Glycophorin C |
|---|---|
| Clone | BRIC10 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000 |
| Applications | Western Blot, Flow Cytometry |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017780
![]() |
Novus Biologicals
NB100665220.025MG |
0.025 mg |
Each for $206.50
|
|
|||||
NB10066522
![]() |
Novus Biologicals
NB10066522 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
Glycophorin C Monoclonal specifically detects Glycophorin C in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
| Glycophorin C | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Cancer, Signal Transduction | |
| P04921 | |
| 2995 | |
| Human erythrocytes | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| BRIC10 | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD236, CD236 antigen, CD236R, GE, GLPC, glycoconnectin, glycophorin C (Gerbich blood group), glycophorin-C, glycophorin-D, Glycoprotein beta, GPCMGC126191, GPD, GYPD, MGC117309, MGC126192, PAS-2, PAS-2', Sialoglycoprotein D | |
| GYPC | |
| IgG1 | |
| Protein G purified | |
| NB100-66522 recognizes an epitope in the N-terminal region of CD236, also known as glycophorin C (GPC). CD236 is an integral membrane protein which carries the Gerbich blood group antigens. Expression of CD236 is not restricted to erythroid cells, but is broadly distributed in both erythroid and non-erythroid tissues. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title