Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glycophorin C Antibody (BRIC100), Novus Biologicals™

Mouse Monoclonal Antibody
$206.00 - $487.50
Specifications
| Antigen | Glycophorin C |
|---|---|
| Clone | BRIC100 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
| Applications | Immunohistochemistry (Frozen), Western Blot, Immunohistochemistry, Immunoprecipitation |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017732
![]() |
Novus Biologicals
NB100655750.025MG |
0.025 mg |
Each for $206.00
|
|
|||||
NB10065575
![]() |
Novus Biologicals
NB10065575 |
0.1 mg |
Each for $487.50
|
|
|||||
Description
Glycophorin C Monoclonal specifically detects Glycophorin C in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.Specifications
| Glycophorin C | |
| 1.0 mg/mL | |
| Immunohistochemistry (Frozen), Western Blot, Immunohistochemistry, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Cancer, Signal Transduction | |
| P04921 | |
| 2995 | |
| Synthetic peptide corresponding to amino acids in the cytoplasmic domain of GPC | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| BRIC100 | |
| Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD236, CD236 antigen, CD236R, GE, GLPC, glycoconnectin, glycophorin C (Gerbich blood group), glycophorin-C, glycophorin-D, Glycoprotein beta, GPCMGC126191, GPD, GYPD, MGC117309, MGC126192, PAS-2, PAS-2', Sialoglycoprotein D | |
| GYPC | |
| IgG1 | |
| Protein A or G purified | |
| Recognizes a 17 amino acid residue within the cytoplasmic domain of glycophorin C (GPC), which has recently been designated CD236. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title