Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glycophorin C Antibody (BRIC100), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10065575AF750
Description
Glycophorin C Monoclonal specifically detects Glycophorin C in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
Glycophorin C | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
GYPC | |
Synthetic peptide corresponding to amino acids in the cytoplasmic domain of GPC | |
Primary | |
Recognizes a 17 amino acid residue within the cytoplasmic domain of glycophorin C (GPC), which has recently been designated CD236. | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen) | |
BRIC100 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen | |
CD236, CD236 antigen, CD236R, GE, GLPC, glycoconnectin, glycophorin C (Gerbich blood group), glycophorin-C, glycophorin-D, Glycoprotein beta, GPCMGC126191, GPD, GYPD, MGC117309, MGC126192, PAS-2, PAS-2′, Sialoglycoprotein D | |
Mouse | |
0.1 mL | |
2995 | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction