Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glycophorin C Antibody (BRIC4), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2333040.025MG
Description
Glycophorin C Monoclonal specifically detects Glycophorin C in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Haemagglutination.Specifications
Glycophorin C | |
Monoclonal | |
0.5 mg/mL | |
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Haemagglutination | |
P04921 | |
GYPC | |
Human erythrocytes | |
0.025 mg | |
Cancer, Signal Transduction | |
2995 | |
Human | |
IgG1 |
Western Blot, Immunohistochemistry, Hemagglutination Assay, Immunohistochemistry (Frozen), Flow Cytometry | |
BRIC4 | |
Unconjugated | |
TBS with 0.09% Sodium Azide | |
CD236, CD236 antigen, CD236R, GE, GLPC, glycoconnectin, glycophorin C (Gerbich blood group), glycophorin-C, glycophorin-D, Glycoprotein beta, GPCMGC126191, GPD, GYPD, MGC117309, MGC126192, PAS-2, PAS-2', Sialoglycoprotein D | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
NBP2-33304 recognises the erythrocyte transmembrane glycoprotein CD236, also known as glycophorin C (GPC). CD236 is an integral membrane protein which carries the Gerbich blood group antigens. Expression of CD236 is not restricted to erythroid cells, but is broadly distributed in both erythroid and non-erythroid tissues. Clone BRIC4 specifically reacts with an epitope located within residues 2-21 of CD236. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction