Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Glycoprotein 78, Rat, Unlabeled, Clone: 3F3A,

Catalog No. MABC949 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABC949 200 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC949 Supplier MilliporeSigma™ Supplier No. MABC949
Add to Cart
Add to Cart

Rat Monoclonal Antibody

E3 ubiquitin-protein ligase AMFR (UniProt Q9UKV5; also known as AMF receptor, Autocrine motility factor receptor, gp78, RING finger protein 45) is encoded by the RNF45 (also known as AMFR) gene (Gene ID 267) in human. Glycoprotein 78 (gp78) is an endoplasmic reticulum (ER) membrane-anchored E3 ubiquitin ligase that plays a key role in ER-associated degradation (ERAD). Gp78 is localized to a mitochondria-associated ER domain, where it induces mitochondrial fragmentation by targeting the mitochondrial fusion proteins mitofusin 1 and 2 (Mfn1 and Mfn2, respectively) for degradation upon mitochondrial depolarization. Other known ERAD substrates processed by gp78 include ApoB lipoprotein, HMG CoA reductase, CD3-delta, cystic fibrosis transmembrane conductance regulator, the metastasis suppressor KAI1, unglycosylated prion protein (PrP), and the T-cell receptor. Gp78 is a 7-transmembrane protein (a.a. 82-102, 122-142, 145-165, 186-206, 215-235, 276-296, 429-449) with a RING-type Zinc finger (a.a. 341-379), a CUE domain (a.a. 456-498), and a VCP/p97-interacting motif (VIM; a.a. 622-640).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glycoprotein 78
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 3F3A
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rat IgM in buffer containing PBS without azide.
Gene RNF45, AMFR
Gene Accession No. Q9UKV5
Host Species Rat
Immunogen Peanut lectin (PNA) resin-purified membrane glycoprotein 78 from B16-F1 cells (Nabi, I.R., and Raz, A. (1987). Int. J. Cancer. 40(3):396-402).
Purification Method Purified by Ion-Exchange Chromatography
Quantity 200 μL
Regulatory Status RUO
Research Discipline Apoptosis & Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_001135
Test Specificity Clone 3F3A detected glycoprotein 78 (gp78) and a ∽150 kDa glycoprotein in B16-F1 mouse melanoma cell extract. Cell extract sialidase treatment decreased the antibody's immunoreactivity toward gp78 and abolished ∽150 kDa band detection (Nabi, I.R., et al. (1990). Cancer Res. 50(2):409-414).
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgM κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.