Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glycoprotein V/CD42d Antibody (1C2), Alexa Fluor™ 647, Novus Biologicals™

Armenian Hamster Monoclonal Antibody
Supplier: Novus Biologicals NBP200071AF647
Description
Glycoprotein V/CD42d Monoclonal specifically detects Glycoprotein V/CD42d in Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
Glycoprotein V/CD42d | |
Monoclonal | |
Alexa Fluor 647 | |
platelet glycoprotein V, CD42d, glycoprotein V (platelet), GPV | |
Armenian Hamster | |
Affinity Purified | |
RUO | |
2814 | |
Store at 4C in the dark. |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
1C2 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
GP5 | |
The immunogen for this antibody is CD42d (platelet glycoprotein V). | |
0.1 mL | |
Primary | |
Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction