Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glycoprotein V/CD42d Antibody (1C2), Alexa Fluor™ 750, Novus Biologicals™

Armenian Hamster Monoclonal Antibody
Supplier: Novus Biologicals NBP200071AF750
Description
Glycoprotein V/CD42d Monoclonal specifically detects Glycoprotein V/CD42d in Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
Glycoprotein V/CD42d | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
GP5 | |
The immunogen for this antibody is CD42d (platelet glycoprotein V). | |
Primary | |
Mouse, Rat | |
IgG |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
1C2 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
platelet glycoprotein V, CD42d, glycoprotein V (platelet), GPV | |
Armenian Hamster | |
0.1 mL | |
2814 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction