Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glycoprotein V/CD42d Antibody (508209), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB4249L
Description
Glycoprotein V/CD42d Monoclonal antibody specifically detects Glycoprotein V/CD42d in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
Glycoprotein V/CD42d | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2814 | |
Store at 4°C in the dark. | |
IgG1 |
Flow Cytometry, CyTOF | |
508209 | |
Flow Cytometry, CyTOF-ready | |
platelet glycoprotein V, CD42d, glycoprotein V (platelet), GPV | |
Mouse myeloma cell line NS0-derived recombinant human Glycoprotein V/CD42d, Gln17-Gly523, Accession # P40197 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction