Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLYCTK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582268
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582268 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582268 Supplier Invitrogen™ Supplier No. PA582268
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IRAAMERAGK QEMLLKPHSR VQVFEGAEDN LPDRDALRAA LAIQQLAEGL TADDLLLVLI SGGGSALLPA PIPPVTLEEK QTLTRLLAAR GATIQELNTI RKALSQLKGG GLAQAAYPAQ VVSLILSDVV G Highest antigen sequence indentity to the following orthologs: Mouse - 94%, Rat - 94%.

This locus encodes a member of the glycerate kinase type-2 family. The encoded enzyme catalyzes the phosphorylation of (R)-glycerate and may be involved in serine degradation and fructose metabolism. Decreased activity of the encoded enzyme may be associated with the disease D-glyceric aciduria. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLYCTK
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GLYCTK
Gene Accession No. Q8IVS8, Q8QZY2
Gene Alias 6230410P16Rik; C430006L12; glycerate kinase; GLYCTK; HBeAg binding protein 4; HBeAg-binding protein 2; HBeAg-binding protein 4; HBeAgBP4A; HBEBP2; HBEBP4; LP5910
Gene Symbols GLYCTK
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GLYCTK. Recombinant protein control fragment (Product #RP-89448).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 132158, 235582
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.