Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GLYCTK Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA528660
Description
Recommended positive controls: A431, HepG2. Predicted reactivity: Bovine (80%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This locus encodes a member of the glycerate kinase type-2 family. The encoded enzyme catalyzes the phosphorylation of (R)-glycerate and may be involved in serine degradation and fructose metabolism. Decreased activity of the encoded enzyme may be associated with the disease D-glyceric aciduria. Alternatively spliced transcript variants have been described.
Specifications
GLYCTK | |
Polyclonal | |
Unconjugated | |
GLYCTK | |
6230410P16Rik; C430006L12; glycerate kinase; GLYCTK; HBeAg binding protein 4; HBeAg-binding protein 2; HBeAg-binding protein 4; HBeAgBP4A; HBEBP2; HBEBP4; LP5910 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
132158 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.54 mg/mL | |
0.1M Tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q8IVS8 | |
GLYCTK | |
Recombinant fragment corresponding to a region within amino acids 249 and 523 of Human GLYCTK. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction