Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glycylsarcosine 97.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
1g
5g
Specifications
Specifications
| Purity | ≥97.0% (HPLC) |
| CAS | 29816-01-1 |
| Color | White |
| Content And Storage | Refrigerator |
| Format | Crystal-Powder at 20°C |
| Molecular Weight (g/mol) | 146.15 |
| Product Type | Glycylsarcosine |
| Quantity | 1g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction