Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glyoxalase I Antibody (043), Alexa Fluor™ 532, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290549AF532
Description
Glyoxalase I Monoclonal specifically detects Glyoxalase I in Mouse samples. It is validated for ELISA.Specifications
Glyoxalase I | |
Monoclonal | |
Alexa Fluor 532 | |
Aldoketomutase, EC 4.4.1.5, GLOD1, Glx I, GLYI, glyoxalase domain containing 1, glyoxalase Ialdoketomutase, Ketone-aldehyde mutase, lactoyl glutathione lyase, lactoylglutathione lyase, methylglyoxalase, S-D-lactoylglutathione methylglyoxal lyase | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Glyoxalase I (NP_079650.3; Ala 2-Ile 184). | |
0.1 mL | |
Cancer, Core ESC Like Genes, Signal Transduction, Stem Cell Markers | |
2739 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
43 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction