Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glyoxalase I Antibody (9F11), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259369MFV610
Description
Glyoxalase I Monoclonal antibody specifically detects Glyoxalase I in Non-species specific samples. It is validated for Flow CytometrySpecifications
Glyoxalase I | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Non-species specific | |
Purified |
Flow Cytometry | |
9F11 | |
Flow Cytometry | |
Aldoketomutase, EC 4.4.1.5, GLOD1, Glx I, GLYI, glyoxalase domain containing 1, glyoxalase Ialdoketomutase, Ketone-aldehyde mutase, lactoyl glutathione lyase, lactoylglutathione lyase, methylglyoxalase, S-D-lactoylglutathione methylglyoxal lyase | |
Synthetic Methylglyoxal modified Keyhole Limpet Hemocyanin (KLH). | |
0.1 mL | |
Cancer, Core ESC Like Genes, Signal Transduction, Stem Cell Markers | |
2739 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction