Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glyoxalase I Antibody (CPTC-GLO1-3), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308556AF750
Description
Glyoxalase I Monoclonal antibody specifically detects Glyoxalase I in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
Glyoxalase I | |
Monoclonal | |
Alexa Fluor 750 | |
Aldoketomutase, EC 4.4.1.5, GLOD1, Glx I, GLYI, glyoxalase domain containing 1, glyoxalase Ialdoketomutase, Ketone-aldehyde mutase, lactoyl glutathione lyase, lactoylglutathione lyase, methylglyoxalase, S-D-lactoylglutathione methylglyoxal lyase | |
Recombinant full-length human Glyoxalase I protein (Uniprot: Q04760) | |
100 μg | |
Cancer, Core ESC Like Genes, Signal Transduction, Stem Cell Markers | |
2739 | |
Store at 4°C in the dark. | |
IgG1 |
Western Blot, Immunohistochemistry (Paraffin) | |
CPTC-GLO1-3 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction