Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFAP Antibody (5C10), Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 14 publications

$257.00 - $596.50

Specifications

Antigen GFAP
Clone 5C10
Concentration 1.0 mg/mL
Dilution Western Blot 1:5000, Simple Western 1:3000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin 1:1000, Immunohistochemistry-Frozen 1:1000, Immunohistochemistry Free-Floating 1:1000
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP105197SS
SDP
View Documents
Novus Biologicals
NBP105197SS
0.025 mL
Each for $257.00
Only null left
Add to Cart
 
NBP105197
SDP
View Documents
Novus Biologicals
NBP105197
0.1 mL
Each for $596.50
Only null left
Add to Cart
 
Description

Description

GFAP Monoclonal specifically detects GFAP in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating.
Specifications

Specifications

GFAP
1.0 mg/mL
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Astrocyte Markers, Cancer, Cell Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Inflammation, Neurodegeneration, Neuroscience, Signal Transduction, Stem Cell Markers
PBS, 50% glycerol (1:1) with 5mM Sodium Azide
2670
This GFAP Antibody (5C10) was developed against a preparation of purified pig spinal cord GFAP
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
50 kDa
5C10
Western Blot 1:5000, Simple Western 1:3000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin 1:1000, Immunohistochemistry-Frozen 1:1000, Immunohistochemistry Free-Floating 1:1000
Monoclonal
Ascites
RUO
Human, Mouse, Rat, Pig, Bovine, Equine
FLJ45472, GFAP astrocytes, glial fibrillary acidic protein
GFAP
IgG1
Immunogen affinity purified
Human, mouse, rat, chicken, bovine, porcine and equine. Reacts with all other mammalian and avian species tested to date.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.