Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glyoxalase II/HAGH Antibody (002), mFluor Violet 500 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290061MFV500
Description
Glyoxalase II/HAGH Monoclonal antibody specifically detects Glyoxalase II/HAGH in Human samples. It is validated for ELISASpecifications
Glyoxalase II/HAGH | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
002 | |
ELISA | |
EC 3.1.2.6, GLO2hydroxyacyl glutathione hydrolase, Glx II, GLXII, Glyoxalase II, HAGH1GLX2, hydroxyacylglutathione hydrolase, hydroxyacylglutathione hydrolase, mitochondrial, hydroxyacylglutathione hydroxylase | |
Rabbit immunized with purified, recombinant Human Glyoxalase II/HAGH (Uniprot#: Q16775-2, Met1-Asp260) | |
0.1 mL | |
Lipid and Metabolism, Vision | |
3029 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction