Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glyoxalase II/HAGH Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299941IR
Description
Glyoxalase II/HAGH Polyclonal specifically detects Glyoxalase II/HAGH in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Glyoxalase II/HAGH | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 755 | |
EC 3.1.2.6, GLO2hydroxyacyl glutathione hydrolase, Glx II, GLXII, Glyoxalase II, HAGH1GLX2, hydroxyacylglutathione hydrolase, hydroxyacylglutathione hydrolase, mitochondrial, hydroxyacylglutathione hydroxylase | |
Produced in rabbits immunized with purified, recombinant Glyoxalase II/HAGH (Q16775-2; Met 1-Asp 260). | |
0.1 mL | |
Lipid and Metabolism, Vision | |
3029 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction