Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glyphosate Chicken anti-Chemical, Polyclonal, ELTI
SDP

Catalog No. 502100671
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-210-0671 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-210-0671 Supplier Vector Laboratories Supplier No. C16P
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chicken Polyclonal Antibody

Glyphosate Polyclonal antibody specifically detects Glyphosate in Chemical samples. It is validated for Affinity Purification, ELISA

Specifications

Antigen Glyphosate
Applications Affinity Purification, ELISA
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide; pH
Host Species Chicken
Immunogen BSA-Glyphosate conjugate
Purification Method PEG precipitation
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgY
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.