Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glypican 1 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP142718C
Description
Glypican 1 Polyclonal specifically detects Glypican 1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
Glypican 1 | |
Polyclonal | |
Western Blot, ELISA, Immunohistochemistry | |
FLJ38078, glypican, glypican 1, glypican proteoglycan 1, glypican-1 | |
Glypican 1 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region of human glypican-1 protein. (Uniprot: P35052) | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, ELISA, Immunohistochemistry | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
2817 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction