Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Glypican 1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140250
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140250 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140250 Supplier Invitrogen™ Supplier No. PA5140250
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: GDGCLDDLCS RKVSRKSSSS RTPLTHALPG LSEQEGQKTS AA Highest antigen sequence indentity to the following orthologs: Rat - 86%, Mouse - 86%.

Cell surface heparan sulfate proteoglycans are composed of a membrane-associated protein core substituted with a variable number of heparan sulfate chains. Members of the glypican-related integral membrane proteoglycan family (GRIPS) contain a core protein anchored to the cytoplasmic membrane via a glycosyl phosphatidylinositol linkage. These proteins may play a role in the control of cell division and growth regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glypican 1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GPC1
Gene Accession No. P35052
Gene Alias AI462976; glypican; glypican 1; glypican proteoglycan 1; glypican-1; Gpc1; GPI-anchored cell surface heparan sulfate proteoglycan; HSPG M12; HSPGM12; Secreted glypican-1
Gene Symbols GPC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GPC1. Recombinant protein control fragment (Product #RP-109211).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2817
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.