Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glypican 3 Antibody (1G12 + GPC3/863), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP247763MFV450
Description
Glypican 3 Monoclonal antibody specifically detects Glypican 3 in Human samples. It is validated for Flow CytometrySpecifications
Glypican 3 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
1G12 + GPC3/863 | |
Flow Cytometry | |
DGSX, glypican 3, glypican proteoglycan 3, glypican-3, GTR2-2, heparan sulphate proteoglycan, Intestinal protein OCI-5, MXR7, OCI5, OCI-5, secreted glypican-3, SGB, SGBS, SGBS1SDYS | |
Recombinant fragment containing amino acids 511-580 of human Glypican 3 (1G12); Recombinant full-length human GPC3 protein (GPC3/863) (Uniprot: P51654) | |
0.1 mL | |
Cancer, Cell Biology, Cytoskeleton Markers, Signal Transduction, Stem Cells | |
2719 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction