Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Glypican-3 (Clone 1G12), Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Glypican-3 (Clone 1G12), Mouse Monoclonal Antibody.

Supplier:  Epredia™ MS1939R7

 View more versions of this product

Catalog No. MS1939R7


Description

Description

Glypican-3 (GPC3), is a member of the glypican family of heparan sulfate proteoglycans.  GPC3 has an important role in cell growth and differentiation. GPC3 expression has been observed in embryonic tissue including fetal liver while benign and healthy liver specimens are negative. Literature suggests that GPC3 is overexperssed in hepatocellular carcinoma (HCC) while it appears to be down-regulated in several other malignancies such as mesothelioma, ovarian carcinoma, breast carcinoma and intrahepatic Cholangiocarcinomas.  Studies also suggest a potential link between GPC3 expression and poor prognosis in HCC3.  In addition, Avidel-Ronen et al suggests that Glypican-3 is a potential marker for early detection of squamous cell carcinoma of the lung2.  

Host Species: Mouse
Clone: 1G12
Isotype: IgG1
Species Reactivity
: Human. Others not tested
Epitope: Not determined
Immunogen: The last 70 amino acids of the core protein
Molecular Weight: ∼60kDa
Positive Control: Hepatocellular carcinoma or placenta
Cellular Localization: Cytoplasmic
Recommended for: Immunohistochemistry
Specifications

Specifications

Immunohistochemistry
Monoclonal
Unconjugated
The last 70 amino acids of the core protein
Hepatocellular carcinoma or placenta
7 mL
Primary
This product contains sodium azide and is stable for 24 months from the date of manufacture when stored at 2-8°C. Do not use after expiration date indicated on label of the product. If reagent is not storedas recommended, performance must be validated by the user
Cytoplasmic
1G12
Mouse
∼60kDa
Unpurified
IVD
Human
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.