Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Glypican 3 Recombinant Rabbit Monoclonal Antibody (7M2X9)
GREENER_CHOICE

Catalog No. PIMA535154
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535154 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535154 Supplier Invitrogen™ Supplier No. MA535154
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Glypican 3 Recombinant Monoclonal Antibody for ELISA, ICC/IF, Western Blot

GPC3 is a cell surface proteoglycan that bears heparan sulfate. This protein may be involved in the suppression/modulation of growth in the predominantly mesodermal tissues and organs, and may play a role in the modulation of IGF2 interactions with its receptor and thereby modulate its function. Members of the glypican-related integral membrane proteoglycan family contain a core protein anchored to the cytoplasmic membrane via a glycosyl phosphatidylinositol (GPI) linkage. These proteins may play a role in the control of cell division, growth regulation, and tumor predisposition. Deletion mutations in GPC3 are the cause of Simpson-Golabi-Behmel syndrome (SGBS), also known as Simpson dysmorphia syndrome (SDYS). SGBS is a condition characterized by pre- and postnatal overgrowth (gigantism) with visceral and skeletal anomalies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glypican 3
Applications ELISA, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 7M2X9
Concentration 0.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.05% BSA and 0.02% sodium azide; pH 7.3
Gene GPC3
Gene Accession No. P51654
Gene Alias defective in Simpson-Golabi-Behmel overgrowth syndrome; DGSX; glypican 3; glypican proteoglycan 3; glypican-3; Glypican-3 alpha subunit; Glypican-3 beta subunit; Gpc3; GTR22; GTR2-2; heparan sulphate proteoglycan; intestinal protein OCI-5; MXR7; OCI5; OCI-5; proteoglycan GPC3; SDYS; secreted glypican-3; SGB; SGBS; SGBS1
Gene Symbols GPC3
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2719
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.