Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Glypican 3 Recombinant Rabbit Monoclonal Antibody (7K5V6)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA551509
Description
Glypican 3 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow, ELISA
GPC3 is a cell surface proteoglycan that bears heparan sulfate. This protein may be involved in the suppression/modulation of growth in the predominantly mesodermal tissues and organs, and may play a role in the modulation of IGF2 interactions with its receptor and thereby modulate its function. Members of the glypican-related integral membrane proteoglycan family contain a core protein anchored to the cytoplasmic membrane via a glycosyl phosphatidylinositol (GPI) linkage. These proteins may play a role in the control of cell division, growth regulation, and tumor predisposition. Deletion mutations in GPC3 are the cause of Simpson-Golabi-Behmel syndrome (SGBS), also known as Simpson dysmorphia syndrome (SDYS). SGBS is a condition characterized by pre- and postnatal overgrowth (gigantism) with visceral and skeletal anomalies.
Specifications
Glypican 3 | |
Recombinant Monoclonal | |
0.76 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
P51654 | |
GPC3 | |
A synthetic peptide corresponding to a sequence within amino acids 481-580 of human Glypican 3 (GPC3) (NP_004475.1). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
ELISA, Flow Cytometry, Western Blot, Immunocytochemistry | |
7K5V6 | |
Unconjugated | |
GPC3 | |
defective in Simpson-Golabi-Behmel overgrowth syndrome; DGSX; glypican 3; glypican proteoglycan 3; glypican-3; Glypican-3 alpha subunit; Glypican-3 beta subunit; Gpc3; GTR22; GTR2-2; heparan sulphate proteoglycan; intestinal protein OCI-5; MXR7; OCI5; OCI-5; proteoglycan GPC3; SDYS; secreted glypican-3; SGB; SGBS; SGBS1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
2719 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction