Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glypican 4 Antibody (51E3), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP249892R
Description
Glypican 4 Monoclonal specifically detects Glypican 4 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Glypican 4 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
GPC4 | |
Recombinant human GPC4(401-529aa) purified from E. coli | |
100 μL | |
Cancer, Stem Cells | |
2239 | |
Human | |
Purified |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
51000 | |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
glypican 4, glypican proteoglycan 4, glypican-4, K-glypicandJ900E8.1 (glypican 4) | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Anti-human GPC4 mAb, clone AT51E3, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human GPC4 protein. | |
Store at 4C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction