Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Glypican 4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588766
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588766 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588766 Supplier Invitrogen™ Supplier No. PA588766
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SESAFSARFR PHHPEERPTT AAGTSLDRLV TDVKEKLKQA KKFWSSLPSN VCNDERMAAG NGNEDDCWNG KGKSRYLFAV TGNGLANQGN NPEVQVDTSK PDILILRQIM ALRVMTSKMK NAYNGNDVDF FDISDESSGE GSGSGCEYQQ CPSEFDYNAT DHAGKSANEK ADSAGVRPGA QAYLLTVFCI LFLVMQREWR; Positive Samples: U-87MG, A-431, 293T; Cellular Location: Cell membrane, Extracellular side, GPI-anchor, Lipid-anchor, extracellular space.

Cell surface heparan sulfate proteoglycans are composed of a membrane-associated protein core substituted with a variable number of heparan sulfate chains. Members of the glypican-related integral membrane proteoglycan family contain a core protein anchored to the cytoplasmic membrane via a glycosyl phosphatidylinositol linkage. These proteins may play a role in the control of cell division and growth regulation. The GPC4 gene is adjacent to the 3' end of GPC3 and may also play a role in Simpson-Golabi-Behmel syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glypican 4
Applications ELISA, Western Blot, Western Blot
Classification Polyclonal
Concentration 0.63 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene GPC4
Gene Accession No. O75487, P51655
Gene Alias 9530073D23Rik; AF311610; AI385680; AI661372; dJ900E8.1 (glypican 4); glypican 4; glypican proteoglycan 4; glypican-4; Gpc4; K-glypican; Secreted glypican-4; UNQ474/PRO937
Gene Symbols GPC4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 357-556 of human GPC4 (NP_0014392).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14735, 2239
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.