Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glypican 5 Antibody (297716), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB2607X
Description
Glypican 5 Monoclonal antibody specifically detects Glypican 5 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
Glypican 5 | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2262 | |
Store at 4°C in the dark. | |
IgG2a |
Western Blot, Flow Cytometry, CyTOF | |
297716 | |
Western Blot, Flow Cytometry, CyTOF-ready | |
bA93M14.1, glypican 5, glypican proteoglycan 5, glypican-5 | |
Mouse myeloma cell line NS0-derived recombinant human Glypican 5, Glu25-Thr554, Accession # P78333 | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction