Learn More
Invitrogen™ Glypican 5 Monoclonal Antibody (1C9)
Mouse Monoclonal Antibody
Supplier:  Invitrogen™ MA518886
Catalog No. PIMA518886
Only null left
Add to Cart
Description
Description
Peptide Sequence: MDAQTWPVGF RCLLLLALVG SARSEGVQTC EEVRKLFQWR LLGAVRGLPD SPRAGPDLQV CISKKPTCCT RKMEERYQIA ARQDMQQFLQ TSSSTLKFLI SRNAAAFQET LETLIKQAEN YTSILFCSTY RNMALEAAAS VQEFFTDVGL YLFGADVNPE EFVNRFFDSL FPLVYNHLIN PGVTDSSLEY SECIRMARRD VSPFGNIPQR VMGQMGRSLL PSRTFLQALN LGIEVINTTD YLHFSKECSR ALLKMQYCPH CQGLALTKPC MGYCLNVMRG CLAHMAELNP HWHAYIRSLE ELSDAMHGTY DIGHVLLNFH LLVNDAVLQA HLNGQKLLEQ VNRICGRPVR TPTQSPRCSF DQSKEKHGMK TTTRNSEETL ANRRKEFINS LRLYRSFYGG LADQLCANEL AAADGLPCWN GEDIVKSYTQ RVVGNGIKAQ SGNPEVKVKG IDPVINQIID KLKHVVQLLQ GRSPKPDKWE LLQLGSGGGM VEQVSGDCDD EDGCGGSGSG EVKRTLKITD WMPDDMNFSD VKQIHQTDTG STLDTTGAGC AVATESMTFT LISVVMLLPG IW.
Cell surface heparan sulfate proteoglycans are composed of a membrane-associated protein core substituted with a variable number of heparan sulfate chains. Members of the glypican-related integral membrane proteoglycan family (GRIPS) contain a core protein anchored to the cytoplasmic membrane via a glycosyl phosphatidylinositol linkage. These proteins may play a role in the control of cell division and growth regulation. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Glypican 5
Monoclonal
0.5 mg/mL
PBS with no preservative; pH 7.4
P78333
GPC5
GPC5 (AAH39730, 1 a.a. ~ 572 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG2a κ
ELISA, Immunoprecipitation
1C9
Unconjugated
GPC5
A230034F01Rik; bA93M14.1; glypican 5; glypican proteoglycan 5; glypican-5; Gpc5; Secreted glypican-5
Mouse
Affinity chromatography
RUO
2262
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.