Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glypican 6 Antibody (348701), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB2845Z
Description
Glypican 6 Monoclonal antibody specifically detects Glypican 6 in Human samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), CyTOFSpecifications
Glypican 6 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
10082 | |
Store at 4°C in the dark. | |
IgG1 |
Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
348701 | |
Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
glypican 6, glypican proteoglycan 6, glypican-6, MGC126288, OMIMD1 | |
Mouse myeloma cell line NS0-derived recombinant human Glypican 6, Asp24-Val527, Accession # Q9Y625 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction