Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Glypican 6 Monoclonal Antibody (348701)
GREENER_CHOICE

Catalog No. PIMA524086
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524086 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524086 Supplier Invitrogen™ Supplier No. MA524086
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

In direct ELISAs and Western blots, no cross-reactivity with recombinant human Glypican 2, 3, or 5 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

The glypicans comprise a family of glycosylphosphatidylinositol-anchored heparan sulfate proteoglycans. The glypicans have been implicated in the control of cell growth and division. Glypican 6 is a putative cell surface coreceptor for growth factors, extracellular matrix proteins, proteases and anti-proteases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glypican 6
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 348701
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Gpc6
Gene Accession No. Q9Y625
Gene Alias glypican 6; glypican proteoglycan 6; glypican-6; GPC6; OMIMD1; Secreted glypican-6; UNQ369/PRO705
Gene Symbols Gpc6
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Glypican 6 Asp24-Val527.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10082
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.