Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Glypican 6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144710
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144710 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144710 Supplier Invitrogen™ Supplier No. PA5144710
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AYNGNDVNFQ DTSDESSGSG SGSGCMDDVC PTEFEFVTTE APAVDPDRRE VDSSAAQRGH SLLSWSLTCI VLAL Antigen sequence identity: Mouse 86%; Rat 86%.

The glypicans comprise a family of glycosylphosphatidylinositol-anchored heparan sulfate proteoglycans. The glypicans have been implicated in the control of cell growth and division. Glypican 6 is a putative cell surface coreceptor for growth factors, extracellular matrix proteins, proteases and anti-proteases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glypican 6
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Gpc6
Gene Accession No. Q9Y625
Gene Alias glypican 6; glypican proteoglycan 6; glypican-6; GPC6; OMIMD1; Secreted glypican-6; UNQ369/PRO705
Gene Symbols Gpc6
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 477-550. Recombinant protein control fragment (Product #RP-110174).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10082
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.