Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLYR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546762
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546762 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546762 Supplier Invitrogen™ Supplier No. PA546762
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: SSHNSSDDKN RRNSSEERSR PNSGDEKRKL SLSEGKVKKN MGEGKKRVSS.

GLYR1 may have oxidoreductase activity. It regulates p38 MAP kinase activity by mediating stress activation of p38alpha/MAPK14 and specifically regulating MAPK14 signaling. It indirectly promotes phosphorylation of MAPK14 and activation of ATF2. The phosphorylation of MAPK14 requires upstream activity of MAP2K4 and MAP2K6. It is recruited on chromatin, recognizes and binds trimethylated 'Lys-36' of histone H3 (H3K36me3).
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLYR1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GLYR1
Gene Accession No. Q49A26
Gene Alias 2810419J22Rik; 3930401k13rik; 3-hydroxyisobutyrate dehydrogenase-like protein; AW545332; BM045; Cytokine-like nuclear factor N-PAC; glyoxylate reductase 1 homolog; glyoxylate reductase 1 homolog (Arabidopsis); GLYR1; HIBDL; hNDF; NDF; NP60; Npac; N-pac; nuclear protein 60 kDa; nuclear protein 60kDa; nuclear protein NP60; Nuclear protein of 60 kDa; Nucleosome-destabilizing factor; Putative oxidoreductase GLYR1; RGD1309459
Gene Symbols GLYR1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human GLYR1 (aa 100-149).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84656
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.