Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GLYR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561690
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561690 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561690 Supplier Invitrogen™ Supplier No. PA561690
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IVNPPKDLKK PRGKKCFFVK FFGTEDHAWI KVEQLKPYHA HKEEMIKINK GKRFQQAVDA VEEFLRRAKG KDQTSSHNSS D Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

GLYR1 may have oxidoreductase activity. It regulates p38 MAP kinase activity by mediating stress activation of p38alpha/MAPK14 and specifically regulating MAPK14 signaling. It indirectly promotes phosphorylation of MAPK14 and activation of ATF2. The phosphorylation of MAPK14 requires upstream activity of MAP2K4 and MAP2K6. It is recruited on chromatin, recognizes and binds trimethylated 'Lys-36' of histone H3 (H3K36me3).
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLYR1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GLYR1
Gene Accession No. Q49A26
Gene Alias 2810419J22Rik; 3930401k13rik; 3-hydroxyisobutyrate dehydrogenase-like protein; AW545332; BM045; Cytokine-like nuclear factor N-PAC; glyoxylate reductase 1 homolog; glyoxylate reductase 1 homolog (Arabidopsis); GLYR1; HIBDL; hNDF; NDF; NP60; Npac; N-pac; nuclear protein 60 kDa; nuclear protein 60kDa; nuclear protein NP60; Nuclear protein of 60 kDa; Nucleosome-destabilizing factor; Putative oxidoreductase GLYR1; RGD1309459
Gene Symbols GLYR1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GLYR1. Recombinant protein control fragment (Product #RP-103018).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84656
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.