Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GlyT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116669
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116669 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116669 Supplier Invitrogen™ Supplier No. PA5116669
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: SH-SY5Y, K562, MCF7, mouse liver, C6, rat liver Immunogen sequence: MSGGDTRAAI ARPRMAAAHG PVAPSSPEQV TLLPVQRSFF LPPFSGATPS TSLAESVLKV WHGAYNSGLL PQLMAQHSLA MAQNGAVPSE ATKRDQNLKR GNWGNQIEFV.

Slc6a9 is a glycine transporter (GlyTs) that belong to a family of high affinity Na+- and Cl-dependent neurotransmitter transporters proteins. There are two subtypes of glycine transporters; GlyT1 (Slc6a9) and GlyT2. Slc6a9 are predominantly located on glia within caudal brain areas. The amino acid glycine acts as an inhibitory neurotransmitter in the central nervous system. The protein encoded by the Slc6a9 gene is one of two transporters that stop glycine signaling by removing it from the synaptic cleft. Diseases associated with SLC6A9 include Glycine Encephalopathy With Normal Serum Glycine and Infantile Glycine Encephalopathy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GlyT1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.50 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene SLC6A9
Gene Accession No. P28571, P28572, P48067
Gene Alias glycine transporter; glycine transporter 1; glycine transporter variant 1a; glycine-1 transporter; GLYT1; glyT-1; GLYT-1b; Slc6a9; Sodium- and chloride-dependent glycine transporter 1; solute carrier family 6 (neurotransmitter transporter, glycine), member 9; solute carrier family 6 member 9
Gene Symbols SLC6A9
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116509, 14664, 6536
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.